Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274448_m1
          See other LINC01133 GE Assays ›
          Gene Symbol:
          LINC01133
          Gene Name:
          long intergenic non-protein coding RNA 1133
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 159961224 - 159979086 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274448_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01133
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038849.1 2 - 3 642 126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB853513.1 - 1 - 2 Chr. 1 - 159976688 - 159978990 [-] on Build GRCh38 126
          GenBankmRNA AA171762.1 - 1 - 2 65 Chr. 1 - 159977039 - 159978927 [+] on Build GRCh38 126
          GenBankmRNA CA445323.1 - 1 - 2 Chr. 1 - 159976901 - 159979061 [-] on Build GRCh38 126
          GenBankmRNA AA174008.1 - 1 - 2 105 Chr. 1 - 159976999 - 159978848 [+] on Build GRCh38 126
          GenBankmRNA AA169419.1 - 1 - 2 93 Chr. 1 - 159977011 - 159978986 [+] on Build GRCh38 126
          GenBankmRNA CA428652.1 - 1 - 2 Chr. 1 - 159976842 - 159978983 [-] on Build GRCh38 126
          GenBankmRNA BX090035.1 - 1 - 2 296 Chr. 1 - 159976806 - 159979018 [+] on Build GRCh38 126
          GenBankmRNA BE392165.1 - 1 - 2 167 Chr. 1 - 159976927 - 159978984 [+] on Build GRCh38 127
          GenBankmRNA BM997633.1 - 1 - 2 Chr. 1 - 159977008 - 159979062 [-] on Build GRCh38 126

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SLAM family member 9 SLAMF9 CD2F-10,
          CD2F10,
          CD84-H1,
          CD84H1,
          SF2001
          -22.4kb Anti-sense Chr. 1 - 159951492 - 159984730 on NCBI Build GRCh38
          Downstream phosphatidylinositol glycan anchor biosynthesis class M PIGM GPI-MT-I 48.6kb Anti-sense Chr. 1 - 160027672 - 160031993 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010276.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001358.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15136042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20602227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00543584 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 62032 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 434817 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 294520 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 637598 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 640505 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 37354 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P828496 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 32568 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P828496 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P00174908 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00174908 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P828496 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 68231 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 17180 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17864874 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00174918 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17864341 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00174905 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00174908 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2259505 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2259505 - -
          Agilent Human 1B Microarray G4111A A_32_P20609 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P17864874 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00174908 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01021272.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01021272.hg.1 NR_038849.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01021272.hg.1 TC01001358.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175365.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175365.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175365.hg.1 TC0100010276.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100175365.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175365.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175366.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175366.hg.1 TC0100010276.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100175366.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000423943.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 ENST00000443364.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 NR_038849.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010276.hg.1 NR_038849.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033216 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033220 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033216 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033220 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0