Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274460_m1
          See other LOC100129027 GE Assays ›
          Gene Symbol:
          LOC100129027
          Gene Name:
          uncharacterized LOC100129027
          Gene Aliases:
          -
          Chromosome Location:
          Chr.21: 45827841 - 45836419 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274460_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129027
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038876.1 2 - 3 181 73
          GenBank mRNA AK123852.1 - 2 - 3 181 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA726376.1 - 2 - 3 181 Chr. 21 - 45830870 - 45836419 [-] on Build GRCh38 73
          GenBankmRNA DA725199.1 - 2 - 3 181 Chr. 21 - 45830862 - 45836419 [-] on Build GRCh38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105372840 LOC105372840 -230.4kb Anti-sense Chr. 21 - 45593664 - 45603056 on NCBI Build GRCh38
          Downstream poly(rC) binding protein 3 PCBP3 ALPHA-CP3 17.3kb Anti-sense Chr. 21 - 45849961 - 45942454 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100008441.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000550.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK123852.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418399 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418383 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418390 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418374 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418391 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418406 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418394 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418375 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418396 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11418415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32081655 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375681 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25895 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24242 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375681 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375688 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3375681 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03597210 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03597210 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 82980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21006622.hg.1 TC21000550.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21006622.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21006622.hg.1 NR_038876.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21006621.hg.1 TC21000550.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21006621.hg.1 NR_038876.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100157189.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100157189.hg.1 TC2100008441.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100157189.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100157190.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100157190.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100157190.hg.1 TC2100008441.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100157190.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100157190.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 ENST00000380008.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 NR_038876.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008441.hg.1 NR_038876.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019010 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019010 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3263423 AK123852.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3263423 NR_038876.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1913304 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 2680332 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_99241 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1913304 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1913304 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1913304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0