Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274477_g1
          See other MIF-AS1 GE Assays ›
          Gene Symbol:
          MIF-AS1
          Gene Name:
          MIF antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.22: 23893710 - 23898930 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274477_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIF-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038911.1 2 - 3 371 82
          GenBank mRNA BC036909.1 - 2 - 3 371 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI830107.1 - 2 - 3 372 Chr. 22 - 23895180 - 23898931 [-] on Build GRCh38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 2 member 11 SLC2A11 GLUT10,
          GLUT11
          -7.6kb Anti-sense Chr. 22 - 23856703 - 23886112 on NCBI Build GRCh38
          Downstream glutathione S-transferase theta-4-like LOC100652871 39.4kb Anti-sense Chr. 22 - 23935477 - 23942861 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008196.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22001485.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P49334 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 20156 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 41890 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3351894 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P49334 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P49334 - -
          Agilent Human 1B Microarray G4111A A_32_P49334 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556316_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR22010218.hg.1 TC22001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22010218.hg.1 NR_038911.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22010219.hg.1 TC22001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22010219.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22010219.hg.1 NR_038911.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22010220.hg.1 TC22001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22010220.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22010220.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000215754.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000465752.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 ENST00000498385.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009231.hg.1 NM_002415.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200158721.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158721.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200158721.hg.1 TC2200008196.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200158721.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200158721.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000406213.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 ENST00000609510.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008196.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200158720.hg.1 NR_038911.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200158720.hg.1 TC2200008196.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200158720.hg.1 NR_038911.1 Refseq
          Human Genome U133 Plus 2.0 Array 1556316_s_at - -
          Human Genome U133 Plus 2.0 Array 217871_s_at - -
          Human Genome U133 Plus 2.0 Array 1556316_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 217871_PM_s_at - -
          Human Genome U133A 2.0 Array 217871_s_at - -
          Human Genome U133A 2.0 Array 217871_PM_s_at - -
          Human Genome U95 Set 895_PM_at - -
          Human Genome U95 Set 895_at - -
          Human Genome U95 Set 86505_PM_at - -
          Human Genome U95 Set 86505_at - -
          Human Genome U133 Set 217871_s_at - -
          Human Genome U133 Set 217871_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 173844 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7ffdcc7b59-b9k6r:80/100.66.134.75:80.
          git-commit: c6f4609997b4833aba2fd3ad806738f0433cd95c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc