Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274493_m1
          See other LOC100507424 GE Assays ›
          Gene Symbol:
          LOC100507424
          Gene Name:
          uncharacterized LOC100507424
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 2836816 - 2859795 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274493_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100507424
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038933.1 3 - 4 611 146
          GenBank mRNA BU947711.1 - 1 - 2 146
          BU947982.1 - 3 - 4 505 146
          U90917.1 - 1 - 2 1040 146
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA R56428.1 - Chr. - - [-] on Build 146
          GenBankmRNA AI421968.1 - 1 - 2 Chr. 12 - 2858189 - 2859784 [-] on Build GRCh38 146
          GenBankmRNA N46841.1 - 1 - 2 Chr. 12 - 2858049 - 2859790 [-] on Build GRCh38 146
          GenBankmRNA AW051891.1 - 1 - 2 Chr. 12 - 2858093 - 2859784 [-] on Build GRCh38 146
          GenBankmRNA BF939913.1 - 1 - 2 Chr. 12 - 2858168 - 2859784 [-] on Build GRCh38 146
          GenBankmRNA HY279372.1 - 1 - 2 Chr. 12 - 2858192 - 2859795 [-] on Build GRCh38 146
          GenBankmRNA BM675114.1 - 1 - 2 Chr. 12 - 2857845 - 2859791 [-] on Build GRCh38 146
          GenBankmRNA BF940242.1 - 1 - 2 Chr. 12 - 2858026 - 2859784 [-] on Build GRCh38 147
          GenBankmRNA AI241505.1 - 1 - 2 Chr. 12 - 2858183 - 2859791 [-] on Build GRCh38 146
          GenBankmRNA CB055016.1 - 1 - 2 Chr. 12 - 2858051 - 2859791 [-] on Build GRCh38 146
          GenBankmRNA AI492431.1 - 1 - 2 Chr. 12 - 2858116 - 2859791 [-] on Build GRCh38 146
          GenBankmRNA EB388812.1 - 1 - 2 590 Chr. 12 - 2857738 - 2859782 [+] on Build GRCh38 146

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized protein ENSP00000372125 LOC101929469 -797.0 Anti-sense Chr. 12 - 2849221 - 2857215 on NCBI Build GRCh38
          Downstream RAD9-HUS1-RAD1 interacting nuclear orphan 1 RHNO1 C12orf32,
          HKMT1188,
          RHINO
          16.5kb Sense Chr. 12 - 2876258 - 2889525 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012572.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000030.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 66887 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 957258 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 236562 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 506930 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 756578 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 41838 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 205921 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 159575 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559499 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559479 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559478 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19438025 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559480 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559496 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559493 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19438000 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02559485 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19438000 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02559480 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12000426.hg.1 ENST00000545509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000426.hg.1 NR_038933.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000426.hg.1 TC12000030.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000540300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000540300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000540300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 TC1200012572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198626.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 TC1200012572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198631.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 TC1200012572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200198629.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000537710.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000540300.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000540300.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000540300.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 ENST00000545509.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 NR_038933.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012572.hg.1 NR_038933.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A048231 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A048231 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0