Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274808_m1
          See other FLJ33581 GE Assays ›
          Gene Symbol:
          FLJ33581
          Gene Name:
          uncharacterized LOC400839
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 24199767 - 24224588 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274808_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ33581
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040102.1 1 - 2 70 57
          GenBank mRNA AK090900.1 - 1 - 2 70 57
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA153363.1 - 1 - 2 70 Chr. 20 - 24199767 - 24211844 [+] on Build GRCh38 57

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream gamma-glutamyltransferase light chain 1 GGTLC1 GGTL6,
          GGTLA3,
          GGTLA4,
          dJ831C21.1,
          dJ831C21.2
          -211.0kb Anti-sense Chr. 20 - 23985053 - 23989134 on NCBI Build GRCh38
          Downstream translation initiation factor IF-2-like LOC105372585 255.5kb Anti-sense Chr. 20 - 24468226 - 24470506 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000006987.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000160.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17181513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122640 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122716 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31766953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11122692 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 771376 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 453069 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 221178 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 810261 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 418419 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 573009 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 11597 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 548194 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 868285 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 788852 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 252336 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 378070 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 385882 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3832488 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60849 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3832488 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3832488 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03502937 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 99649 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03502927 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21104947 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03502937 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03502937 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03502938 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002015.hg.1 TC20000160.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002015.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002015.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002013.hg.1 TC20000160.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002013.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002013.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002012.hg.1 TC20000160.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002012.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002012.hg.1 NR_040102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002014.hg.1 TC20000160.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002014.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002014.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147706.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147706.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147706.hg.1 TC2000006987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000147706.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147706.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147707.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147707.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147707.hg.1 TC2000006987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000147707.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147707.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147704.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147704.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147704.hg.1 TC2000006987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000147704.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147704.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006987.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147705.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147705.hg.1 NR_040102.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147705.hg.1 TC2000006987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000147705.hg.1 ENST00000457274.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147705.hg.1 NR_040102.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018144 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018144 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0