Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036272.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036272.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036272.hg.1 |
TC03002049.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036273.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036273.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036273.hg.1 |
TC03002049.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013318.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199208.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199208.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199208.hg.1 |
TC0300013318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300199208.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199208.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199209.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199209.hg.1 |
NR_040105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300199209.hg.1 |
TC0300013318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300199209.hg.1 |
ENST00000496220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300199209.hg.1 |
NR_040105.1 |
Refseq |