Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274883_sH
          See other CTAGE7P GE Assays ›
          Gene Symbol:
          CTAGE7P
          Gene Name:
          CTAGE family member 7, pseudogene
          Gene Aliases:
          CTAGE7, CTAGEP, bA500G10.2, rcCTAGE5
          Chromosome Location:
          Chr.10: 130106009 - 130108838 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274883_sH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTAGE7P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_044994.1 1 2420 81
          GenBank mRNA BC036527.1 - 1 2423 81

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream long intergenic non-protein coding RNA 959 LINC00959 Anti-sense Chr. 10 - 130063896 - 130110817 on NCBI Build GRCh38
          Downstream uncharacterized LOC102724883 LOC102724883 2.4kb Sense Chr. 10 - 130111164 - 130127411 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009343.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000931.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16397978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27967609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27967600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27967611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07577204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27967608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27967603 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P348936 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 21335 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P348936 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P348936 - -
          Agilent Human 1B Microarray G4111A A_32_P305657 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7931392 - -
          Affymetrix Human Gene 1.0 ST Array 7931390 - -
          Affymetrix Human Gene 1.0 ST Array 7931391 - -
          Affymetrix Human Gene 1.1 ST Array 7931389 - -
          Affymetrix Human Transcriptome Array 2.0 PSR10011439.hg.1 ENST00000417351.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10011439.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10011439.hg.1 TC10000931.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163911.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163911.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163911.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163911.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163911.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163912.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163912.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163912.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163909.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163909.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163909.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163909.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163909.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163910.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163910.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163910.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163910.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163910.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay TC1000009343.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163906.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163906.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163906.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163908.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163908.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163908.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163908.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163908.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163907.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163907.hg.1 NR_044994.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000163907.hg.1 TC1000009343.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163907.hg.1 ENST00000435712.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163907.hg.1 NR_044994.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 202069 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0