Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04275782_s1
          See other LOC100287015 GE Assays ›
          Gene Symbol:
          LOC100287015
          Gene Name:
          uncharacterized LOC100287015
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 6403556 - 6406548 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04275782_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100287015
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040040.1 2 2320 79
          GenBank mRNA AK025595.1 - 2 2318 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CUB and Sushi multiple domains 1 CSMD1 PPP1R24 -1.4mb Sense Chr. 8 - 2935353 - 4995035 on NCBI Build GRCh38
          Downstream microcephalin 1 MCPH1 BRIT1,
          MCT
          1.5kb Anti-sense Chr. 8 - 6406592 - 6648505 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009415.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000901.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05938268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26211736 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 621572 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16370236 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2763370 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2763370 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556301_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR08011836.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08011836.hg.1 NR_040040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08011836.hg.1 TC08000901.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08011835.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08011835.hg.1 NR_040040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08011835.hg.1 TC08000901.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08011833.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08011833.hg.1 NR_040040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08011833.hg.1 TC08000901.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 TC0800009415.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164455.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164449.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164449.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164449.hg.1 TC0800009415.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164449.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164449.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164451.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164451.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164451.hg.1 TC0800009415.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164451.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164451.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 TC0800009415.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800164456.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000500118.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000523225.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 ENST00000606853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 NR_040040.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009415.hg.1 NR_040040.1 Refseq
          Human Genome U133 Plus 2.0 Array 233174_PM_at - -
          Human Genome U133 Plus 2.0 Array 233174_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233174_at8 - -
          Human Genome U95 Set 83672_at - -
          Human Genome U95 Set 83672_PM_at - -
          Human Genome U133 Set 233174_at - -
          Human Genome U133 Set 233174_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035463 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035463 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 347199 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0