Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04275840_s1
          See other RCAN3AS GE Assays ›
          Gene Symbol:
          RCAN3AS
          Gene Name:
          RCAN3 antisense
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 24496333 - 24502360 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04275840_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RCAN3AS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045407.1 2 313 97
          NR_045408.1 2 354 97
          NR_045409.1 2 261 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG205013.1 - 1 Chr. 1 - 24496266 - 24496584 [-] on Build GRCh38 97
          GenBankmRNA BE551149.1 - 2 321 Chr. 1 - 24496308 - 24499784 [-] on Build GRCh38 97
          GenBankmRNA BG182262.1 - 1 191 Chr. 1 - 24496270 - 24496584 [-] on Build GRCh38 97
          GenBankmRNA BG187486.1 - 1 Chr. 1 - 24496270 - 24496595 [-] on Build GRCh38 97
          GenBankmRNA BM457392.1 - 3 421 Chr. 1 - 24496270 - 24535400 [-] on Build GRCh38 97
          GenBankmRNA BG184860.1 - 1 233 Chr. 1 - 24496270 - 24496584 [-] on Build GRCh38 97
          GenBankmRNA BG202978.1 - 1 193 Chr. 1 - 24496261 - 24496593 [-] on Build GRCh38 97
          GenBankmRNA BG183300.1 - 1 192 Chr. 1 - 24496273 - 24496584 [-] on Build GRCh38 97
          GenBankmRNA BE551182.1 - 2 321 Chr. 1 - 24496308 - 24499784 [-] on Build GRCh38 97
          GenBankmRNA BG183301.1 - 1 234 Chr. 1 - 24496273 - 24496584 [-] on Build GRCh38 97
          GenBankmRNA AW339253.1 - 3 265 Chr. 1 - 24496282 - 24502364 [-] on Build GRCh38 97
          GenBankmRNA BG196301.1 - 1 191 Chr. 1 - 24496273 - 24496598 [-] on Build GRCh38 97
          GenBankmRNA AW134996.1 - 1 Chr. 1 - 24496254 - 24499686 [+] on Build GRCh38 97
          GenBankmRNA CD686666.1 - 1 Chr. 1 - 24495882 - 24496489 [+] on Build GRCh38 97
          GenBankmRNA BG190183.1 - 1 193 Chr. 1 - 24496273 - 24496595 [-] on Build GRCh38 97
          GenBankmRNA CR741819.1 - 2 439 Chr. 1 - 24496270 - 24499902 [-] on Build GRCh38 97
          GenBankmRNA BQ025862.1 - 1 Chr. 1 - 24496261 - 24499771 [+] on Build GRCh38 97
          GenBankmRNA BG194747.1 - 1 191 Chr. 1 - 24496273 - 24496599 [-] on Build GRCh38 97
          GenBankmRNA BG149353.1 - 2 223 Chr. 1 - 24496265 - 24499686 [-] on Build GRCh38 97
          GenBankmRNA BG191673.1 - 1 191 Chr. 1 - 24496270 - 24496598 [-] on Build GRCh38 97
          GenBankmRNA BG195796.1 - 1 191 Chr. 1 - 24496254 - 24496584 [-] on Build GRCh38 97
          GenBankmRNA BF448186.1 - 3 284 Chr. 1 - 24496308 - 24502382 [-] on Build GRCh38 97
          GenBankmRNA BG203977.1 - 1 Chr. 1 - 24496270 - 24496598 [-] on Build GRCh38 97
          GenBankmRNA AA992107.1 - 1 Chr. 1 - 24496270 - 24496598 [+] on Build GRCh38 97
          GenBankmRNA BG194748.1 - 1 191 Chr. 1 - 24496261 - 24496584 [-] on Build GRCh38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream NIPA like domain containing 3 NIPAL3 DJ462O23.2,
          NPAL3
          -23.4kb Anti-sense Chr. 1 - 24413360 - 24472983 on NCBI Build GRCh38
          Downstream RCAN family member 3 RCAN3 DSCR1L2,
          MCIP3,
          RCN3,
          hRCN3
          5.8kb Anti-sense Chr. 1 - 24502351 - 24537020 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0