Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04331453_g1
          See other LINC01348 GE Assays ›
          Gene Symbol:
          LINC01348
          Gene Name:
          long intergenic non-protein coding RNA 1348
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 235065474 - 235069818 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04331453_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01348
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027454.2 2 - 3 620 99
          GenBank mRNA DA457760.1 - 2 - 3 437 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG568342.1 - 2 - 3 558 Chr. 1 - 235067895 - 235069796 [-] on Build GRCh38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101927851 LOC101927851 -103.7kb Sense Chr. 1 - 234957342 - 234963999 on NCBI Build GRCh38
          Downstream translocase of outer mitochondrial membrane 20 TOMM20 MAS20,
          MOM19,
          TOM20
          40.8kb Sense Chr. 1 - 235109341 - 235128941 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100017812.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3407356 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5171 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5172 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40670 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3407356 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3407356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 241533_PM_at - -
          Affymetrix Human Clariom D Assay PSR0100234363.hg.1 ENST00000628622.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100234363.hg.1 NR_027454.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100234363.hg.1 TC0100017812.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100234363.hg.1 ENST00000628622.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100234363.hg.1 NR_027454.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100234360.hg.1 ENST00000628622.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100234360.hg.1 NR_027454.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100234360.hg.1 TC0100017812.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100234360.hg.1 ENST00000628622.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100234360.hg.1 NR_027454.2 Refseq
          Human Genome U133 Plus 2.0 Array 241533_PM_at - -
          Human Genome U133 Plus 2.0 Array 241533_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 241533_at2 - -
          Human Genome U133 Set 241533_PM_at - -
          Human Genome U133 Set 241533_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3301155 NR_027454.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3301155 DA457760.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix