Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04331643_s1
          See other MIR1253 GE Assays ›
          Gene Symbol:
          MIR1253
          Gene Name:
          microRNA 1253
          Gene Aliases:
          MIRN1253, hsa-mir-1253
          Chromosome Location:
          Chr.17: 2748078 - 2748182 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04331643_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR1253
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_031654.1 1 57 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105371592 LOC105371592 -34.9kb Anti-sense Chr. 17 - 2712309 - 2712833 on NCBI Build GRCh38
          Downstream RAP1 GTPase activating protein 2 RAP1GAP2 GARNL4,
          RAP1GA3
          7.5kb Anti-sense Chr. 17 - 2755698 - 3037741 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC1700009457.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001000.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744434 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744434 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16935855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30744429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10195257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700009457.hg.1 ENST00000408273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009457.hg.1 NR_031654.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006566.hg.1 ENST00000609567.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17013348.hg.1 ENST00000408273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17013348.hg.1 NR_031654.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17013348.hg.1 TC17001000.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700006566.hg.1 ENST00000609567.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700172395.hg.1 ENST00000408273.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700172395.hg.1 NR_031654.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700172395.hg.1 TC1700009457.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700172395.hg.1 NR_031654.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700172395.hg.1 ENST00000408273.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009457.hg.1 ENST00000408273.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009457.hg.1 NR_031654.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308828 NR_031654.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308828 NR_031654.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308828 NR_031654.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308828 NR_031654.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0