Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332067_m1
          See other LOC283214 GE Assays ›
          Gene Symbol:
          LOC283214
          Gene Name:
          uncharacterized LOC283214
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 75758455 - 75768647 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332067_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC283214
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046090.1 3 - 4 1311 90
          GenBank mRNA BC039351.1 - 3 - 4 1313 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream monoacylglycerol O-acyltransferase 2 MOGAT2 DGAT2L5,
          DGAT2L5.,
          MGAT2,
          hDC5
          -27.2kb Anti-sense Chr. 11 - 75717819 - 75732958 on NCBI Build GRCh38
          Downstream diacylglycerol O-acyltransferase 2 DGAT2 ARAT,
          GS1999FULL,
          HMFN1045
          6.2kb Anti-sense Chr. 11 - 75768733 - 75801536 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100011663.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11002101.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07881964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07881967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07881968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28300531 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 70341 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 571625 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 826103 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 640953 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 816687 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3690224 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 34136 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3690224 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02471662 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 94472 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11144610 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02471662 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02471660 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11147677 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02471655 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02471662 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561491_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11025176.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11025176.hg.1 NR_046090.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11025176.hg.1 TC11002101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11025175.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11025175.hg.1 NR_046090.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11025175.hg.1 TC11002101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190054.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190054.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190054.hg.1 TC1100011663.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190054.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190054.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190059.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190059.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190059.hg.1 TC1100011663.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190059.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190059.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 ENST00000499390.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 NR_046090.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011663.hg.1 NR_046090.1 Refseq
          Human Genome U133 Plus 2.0 Array 1561491_at - -
          Human Genome U133 Plus 2.0 Array 1561491_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561491_at11 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix