Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332068_m1
          See other LINC00864 GE Assays ›
          Gene Symbol:
          LINC00864
          Gene Name:
          long intergenic non-protein coding RNA 864
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 87396574 - 87407700 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332068_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00864
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046091.1 1 - 2 198 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG400130.1 - 199 Chr. - - [-] on Build 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream NUT family member 2D NUTM2D FAM22D -27.3kb Anti-sense Chr. 10 - 87357720 - 87370695 on NCBI Build GRCh38
          Downstream multiple inositol-polyphosphate phosphatase 1 MINPP1 HIPER1,
          MINPP2,
          MIPP
          95.1kb Anti-sense Chr. 10 - 87502792 - 87553461 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011339.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001488.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27762844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07373988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16369657 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 826951 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 902012 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 589512 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 34003 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 201579 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 396410 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 229965 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3620881 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P12355 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 33389 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27820 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3620881 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P12355 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P02310535 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02310535 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P12355 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 168399 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 100396 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 30877 - -
          Agilent Human 1B Microarray G4111A A_32_P12355 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10018854.hg.1 ENST00000429129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10018854.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10018854.hg.1 TC10001488.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10018856.hg.1 ENST00000429129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10018856.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10018856.hg.1 TC10001488.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10018855.hg.1 ENST00000429129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10018855.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10018855.hg.1 TC10001488.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10018853.hg.1 ENST00000429129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10018853.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10018853.hg.1 TC10001488.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10018852.hg.1 NR_046091.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10018852.hg.1 TC10001488.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000456034.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000456034.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582385.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582385.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011339.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177114.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177114.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177114.hg.1 TC1000011339.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177114.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177114.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177119.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177119.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177119.hg.1 TC1000011339.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177119.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177119.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177116.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177116.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177116.hg.1 TC1000011339.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177116.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177116.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 TC1000011339.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177117.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000439776.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000456034.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000456034.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582385.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582385.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010557.hg.1 ENST00000582543.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177113.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177113.hg.1 TC1000011339.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177113.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 NR_046091.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 TC1000011339.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 ENST00000429129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 ENST00000447936.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177115.hg.1 NR_046091.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 101695 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-54d969bb78-8h2jf:80/100.66.130.62:80.
          git-commit: 3e69e100148d4b3be95fd74bb38be07b481340d5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0