Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000566956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000566956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
ENST00000566956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046290.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046290.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009887.hg.1 |
NR_046290.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
TC1600009887.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176081.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000561547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000566956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000566956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
ENST00000566956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046290.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046290.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009887.hg.1 |
NR_046290.1 |
Refseq |