Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
TC0300011813.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187787.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
TC0300011813.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300187785.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000461931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000475816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
ENST00000488132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011813.hg.1 |
NR_046683.1 |
Refseq |