Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_047467.2 | 2 - 3 | 232 | 87 | ||
GenBank mRNA | DB339345.1 | - | 2 - 3 | 87 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105378430 | LOC105378430 | -6.1kb | Sense | Chr. 10 - 91033166 - 91041255 on NCBI Build GRCh38 | |
Downstream | nudix hydrolase 9 pseudogene 1 | NUDT9P1 |
C10orf98, bA56M3.1 |
104.0kb | Anti-sense | Chr. 10 - 91151995 - 91153083 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() |