Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332603_m1
          See other SNX17 GE Assays ›
          Gene Symbol:
          SNX17
          Gene Name:
          sorting nexin 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 27370496 - 27377533 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332603_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNX17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_049782.1 2 - 3 511 71
          GenBank mRNA AK297054.1 - 2 - 3 395 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DC304856.1 - 2 - 3 391 Chr. 2 - 27370617 - 27373303 [+] on Build GRCh38 71
          GenBankmRNA DC420638.1 - 2 - 3 387 Chr. 2 - 27370620 - 27373309 [+] on Build GRCh38 71
          GenBankmRNA DC359169.1 - 2 - 3 392 Chr. 2 - 27370615 - 27373304 [+] on Build GRCh38 71
          GenBankmRNA CN307078.1 - 2 - 3 324 Chr. 2 - 27370683 - 27374148 [+] on Build GRCh38 71
          GenBankmRNA DC314256.1 - 2 - 3 394 Chr. 2 - 27370614 - 27372734 [+] on Build GRCh38 71
          GenBankmRNA DC388850.1 - 2 - 3 392 Chr. 2 - 27370615 - 27373301 [+] on Build GRCh38 71
          GenBankmRNA DC314369.1 - 2 - 3 389 Chr. 2 - 27370620 - 27373304 [+] on Build GRCh38 71
          GenBankmRNA CF131524.1 - 2 - 3 356 Chr. 2 - 27370651 - 27374159 [+] on Build GRCh38 71
          GenBankmRNA DC339917.1 - 2 - 3 369 Chr. 2 - 27370638 - 27372705 [+] on Build GRCh38 71
          GenBankmRNA DC348555.1 - 2 - 3 403 Chr. 2 - 27370604 - 27373288 [+] on Build GRCh38 71
          GenBankmRNA DC381227.1 - 2 - 3 373 Chr. 2 - 27370636 - 27373311 [+] on Build GRCh38 71
          GenBankmRNA DC297236.1 - 2 - 3 398 Chr. 2 - 27370612 - 27373305 [+] on Build GRCh38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream eukaryotic translation initiation factor 2B subunit delta EIF2B4 EIF-2B,
          EIF2B,
          EIF2Bdelta
          -812.0 Anti-sense Chr. 2 - 27364352 - 27370457 on NCBI Build GRCh38
          Downstream zinc finger protein 513 ZNF513 HMFT0656,
          RP58
          4.5kb Anti-sense Chr. 2 - 27377231 - 27380866 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200007064.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK297054.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cardiac septum development
          cholesterol catabolic process
          intracellular protein transport
          receptor-mediated endocytosis
          signal transduction
          endosomal transport
          regulation of endocytosis
          endocytic recycling
          aorta development
          coronary vasculature development
          signaling receptor binding
          protein binding
          protein C-terminus binding
          phosphatidylinositol binding
          low-density lipoprotein particle receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 ENST00000440760.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 ENST00000484886.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 NR_049782.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 NR_049785.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 TC0200007064.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 ENST00000440760.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 ENST00000484886.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 NR_049782.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200148432.hg.1 NR_049785.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0