Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
TC0X00009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163462.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163464.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163464.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163464.hg.1 |
TC0X00009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00163464.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163464.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163469.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163469.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163469.hg.1 |
TC0X00009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00163469.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163469.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163460.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163460.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163460.hg.1 |
TC0X00009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00163460.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163460.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163468.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163468.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163468.hg.1 |
TC0X00009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00163468.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163468.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
TC0X00009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163466.hg.1 |
NR_073010.2 |
Refseq |