Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04333177_s1
          See other LOC100130744 GE Assays ›
          Gene Symbol:
          LOC100130744 OTULIN
          Gene Name:
          uncharacterized LOC100130744
          OTU deubiquitinase with linear linkage specificity
          Gene Aliases:
          -
          FAM105B, GUM
          Chromosome Location:
          Chr.5: 14712694 - 14716529 on Build GRCh38
          Chromosome Location:
          Chr.5: 14660794 - 14716552 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04333177_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100130744
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046285.1 1 2013 66
          GenBank mRNA AF289590.1 - 1 2033 66
          Gene Symbol: OTULIN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011514151.2 XP_011512453.1 8 7122 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX507119.1 - 1 75 Chr. 5 - 14714632 - 14715242 [+] on Build GRCh38 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ANKH inorganic pyrophosphate transport regulator ANKH ANK,
          CCAL2,
          CMDJ,
          CPPDD,
          HANK,
          MANK
          Anti-sense Chr. 5 - 14704800 - 14871778 on NCBI Build GRCh38
          Downstream microRNA 4637 MIR4637 110.7kb Anti-sense Chr. 5 - 14825929 - 14826012 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500006823.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF289590.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          sprouting angiogenesis
          regulation of tumor necrosis factor-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          innate immune response
          negative regulation of inflammatory response
          canonical Wnt signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          protein linear deubiquitination
          thiol-dependent deubiquitinase
          protein binding
          cysteine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P298495 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P298495 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15118 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 32540 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P298495 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001720249_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001724061_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001721727_at - -
          Affymetrix Human Gene 1.0 ST Array 8104591 - -
          Affymetrix Human Gene 1.1 ST Array 8104590 - -
          Affymetrix HT HG-U133+ PM Array Plate 236228_PM_at - -
          Affymetrix Human Clariom D Assay PSR0500146545.hg.1 NR_046285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500146545.hg.1 TC0500006823.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500146545.hg.1 NR_046285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500146546.hg.1 NR_046285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500146546.hg.1 TC0500006823.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500146546.hg.1 NR_046285.1 Refseq
          Human Genome U133 Plus 2.0 Array 236228_PM_at - -
          Human Genome U133 Plus 2.0 Array 236228_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236228_at3 - -
          Human Genome U95 Set 75558_at - -
          Human Genome U95 Set 75558_PM_at - -
          Human Genome U133 Set 236228_PM_at - -
          Human Genome U133 Set 236228_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 220476 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0