Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008299.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
TC1300008299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156054.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156060.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156060.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156060.hg.1 |
TC1300008299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156060.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156060.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
ENST00000413124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
ENST00000624644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
NR_047040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008299.hg.1 |
NR_047040.1 |
Refseq |