Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
TC0500010371.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172173.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
TC0500010371.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172175.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000506492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
ENST00000507876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010371.hg.1 |
NR_104628.1 |
Refseq |