Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000526546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000526546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000526546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
ENST00000610433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007876.hg.1 |
NR_106806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
ENST00000610433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
NR_106806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
TC1100007876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
NR_106806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156128.hg.1 |
ENST00000610433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000333449.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000526546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000526546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000526546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000532586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000533861.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
ENST00000610433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NM_199337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007876.hg.1 |
NR_106806.1 |
Refseq |