Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04403263_m1
          See other TYW5 GE Assays ›
          Gene Symbol:
          TYW5
          Gene Name:
          tRNA-yW synthesizing protein 5
          Gene Aliases:
          C2orf60, hTYW5
          Chromosome Location:
          Chr.2: 199928911 - 199955736 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04403263_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TYW5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004862.1 2 - 3 472 100
          NR_109906.1 2 - 3 472 100
          GenBank mRNA AK293064.1 - 2 - 3 239 100
          BC025758.1 - 2 - 3 219 100
          BX647677.1 - 2 - 3 473 100
          CN261486.1 - 2 - 3 204 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA021635.1 - 2 - 3 217 Chr. 2 - 199939979 - 199955480 [-] on Build GRCh38 100
          GenBankmRNA DB240040.1 - 1 - 2 91 Chr. 2 - 199933294 - 199948435 [-] on Build GRCh38 100
          GenBankmRNA BG742250.1 - 2 - 3 154 Chr. 2 - 199933117 - 199955417 [-] on Build GRCh38 101
          GenBankmRNA DA574354.1 - 2 - 3 196 Chr. 2 - 199936404 - 199955460 [-] on Build GRCh38 100
          GenBankmRNA DA215655.1 - 2 - 3 246 Chr. 2 - 199936446 - 199955510 [-] on Build GRCh38 100
          GenBankmRNA BX107886.1 - 2 - 3 188 Chr. 2 - 199938749 - 199955452 [-] on Build GRCh38 100
          GenBankmRNA T81742.1 - 189 Chr. - - [-] on Build 100
          GenBankmRNA DC410387.1 - 2 - 3 236 Chr. 2 - 199936002 - 199955500 [-] on Build GRCh38 100
          GenBankmRNA AL530644.3 - 2 - 3 222 Chr. 2 - 199932944 - 199955475 [-] on Build GRCh38 100
          GenBankmRNA DC422110.1 - 2 - 3 239 Chr. 2 - 199936005 - 199955503 [-] on Build GRCh38 100
          GenBankmRNA DA013732.1 - 2 - 3 253 Chr. 2 - 199936430 - 199955516 [-] on Build GRCh38 100
          GenBankmRNA BI769592.1 - 2 - 3 219 Chr. 2 - 199933074 - 199955483 [-] on Build GRCh38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chromosome 2 open reading frame 69 C2orf69 -15.5kb Anti-sense Chr. 2 - 199911256 - 199928273 on NCBI Build GRCh38
          Downstream chromosome 2 open reading frame 47 C2orf47 6.8kb Anti-sense Chr. 2 - 199955317 - 199964125 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA modification
          wybutosine biosynthetic process
          tRNA binding
          iron ion binding
          2-oxoglutarate-dependent dioxygenase activity
          protein homodimerization activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0