Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
ENST00000481625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
ENST00000481625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009042.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
ENST00000481625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
ENST00000481625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009042.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165459.hg.1 |
ENST00000405369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165459.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165459.hg.1 |
TC2200009042.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165459.hg.1 |
NR_109967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165459.hg.1 |
ENST00000405369.3 |
Ensembl |