Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152577.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152577.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152577.hg.1 |
TC0700007590.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700152577.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152577.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152575.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152575.hg.1 |
TC0700007590.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700152575.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007590.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152576.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152576.hg.1 |
NR_110040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152576.hg.1 |
TC0700007590.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700152576.hg.1 |
ENST00000439413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152576.hg.1 |
NR_110040.1 |
Refseq |