Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
ENST00000429872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
ENST00000429872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006464.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
ENST00000429872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
ENST00000429872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006464.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700144242.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700144242.hg.1 |
TC0700006464.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700144242.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
ENST00000429872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
NR_110055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
TC0700006464.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
ENST00000429872.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
NR_110054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700144243.hg.1 |
NR_110055.1 |
Refseq |