Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000453564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000453564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000453564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
TC0700006843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147276.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
TC0700006843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147275.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000446399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000453564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000453564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000453564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
ENST00000455373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006843.hg.1 |
NR_110117.1 |
Refseq |