Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153060.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153060.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153060.hg.1 |
TC0200007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200153060.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153060.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007677.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153059.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153059.hg.1 |
NR_110219.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153059.hg.1 |
TC0200007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200153059.hg.1 |
ENST00000409590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153059.hg.1 |
NR_110219.1 |
Refseq |