Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000423074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008899.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
TC2000008899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162032.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
NR_110623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
TC2000008899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
ENST00000439444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
ENST00000443171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162031.hg.1 |
NR_110623.1 |
Refseq |