Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011717.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
TC0400011717.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180553.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
NR_110660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
TC0400011717.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
ENST00000503266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
ENST00000512219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180552.hg.1 |
NR_110660.1 |
Refseq |