Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200353.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200353.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200353.hg.1 |
TC0100013519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100200353.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200353.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
ENST00000445180.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013519.hg.1 |
NR_110790.1 |
Refseq |