Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000451016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000451016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
ENST00000451016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000418409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000429396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000451016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000451016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
ENST00000451016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013613.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
NR_111922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
TC0700013613.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
ENST00000440504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201815.hg.1 |
NR_111922.1 |
Refseq |