Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
ENST00000571340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007882.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157466.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157466.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157466.hg.1 |
TC1600007882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157466.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157466.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
ENST00000571340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007882.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
TC1600007882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157470.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
NR_119378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
TC1600007882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
ENST00000565189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
ENST00000566383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157472.hg.1 |
NR_119378.1 |
Refseq |