Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007084.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007084.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149690.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149691.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149692.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149692.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149692.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149692.hg.1 |
ENST00000540733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149692.hg.1 |
NR_120473.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149694.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149694.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149694.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149694.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149694.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149696.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149696.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149696.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149696.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149696.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149695.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149695.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149695.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149695.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149695.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149698.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149698.hg.1 |
NR_120472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149698.hg.1 |
TC1200007084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200149698.hg.1 |
ENST00000539583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149698.hg.1 |
NR_120472.1 |
Refseq |