Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
ENST00000534002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166136.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166136.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166136.hg.1 |
TC1100008958.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166136.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166136.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000525797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
ENST00000534002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008958.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166137.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166137.hg.1 |
NR_120552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166137.hg.1 |
TC1100008958.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166137.hg.1 |
ENST00000526101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166137.hg.1 |
NR_120552.1 |
Refseq |