Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04406381_m1
          See other LOC101927055 GE Assays ›
          Gene Symbol:
          LOC101927055
          Gene Name:
          uncharacterized LOC101927055
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 178776926 - 178779963 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04406381_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC101927055
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_120594.1 1 - 2 1826 97
          GenBank mRNA AK123298.1 - 1 - 2 1826 97
          AW444666.1 - 1 - 2 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB305534.1 - 1 - 2 Chr. 2 - 178778354 - 178779957 [-] on Build GRCh38 97
          GenBankmRNA DB302820.1 - 1 - 2 Chr. 2 - 178778488 - 178779962 [-] on Build GRCh38 97
          GenBankmRNA DB308778.1 - 1 - 2 Chr. 2 - 178778314 - 178779962 [-] on Build GRCh38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TTN antisense RNA 1 TTN-AS1 -156.7kb Sense Chr. 2 - 178522827 - 178620217 on NCBI Build GRCh38
          Downstream coiled-coil domain containing 141 CCDC141 CAMDI 49.8kb Anti-sense Chr. 2 - 178825308 - 179050134 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016573.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000631319.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000631319.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 ENST00000631319.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 TC0200016573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224365.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224366.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224366.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224366.hg.1 TC0200016573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200224366.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224366.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 TC0200016573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224368.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 TC0200016573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224367.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224364.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224364.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224364.hg.1 TC0200016573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200224364.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224364.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000578746.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582038.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000582847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000584485.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000590773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000610005.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000627661.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000629094.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000631319.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000631319.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 ENST00000631319.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 NR_120594.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016573.hg.1 NR_120594.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix