Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178996.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178996.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178996.hg.1 |
TC1000011587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178996.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178996.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178997.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178997.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178997.hg.1 |
TC1000011587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178997.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178997.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178998.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178998.hg.1 |
NR_120618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178998.hg.1 |
TC1000011587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178998.hg.1 |
ENST00000548010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178998.hg.1 |
NR_120618.1 |
Refseq |