Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168690.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168690.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168690.hg.1 |
TC1000010083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000168690.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168690.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
ENST00000452911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010083.hg.1 |
NR_120650.1 |
Refseq |