Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008292.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008292.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008292.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008292.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008292.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008292.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156214.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156214.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156214.hg.1 |
TC1000008292.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000156214.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156214.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156216.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156216.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156216.hg.1 |
TC1000008292.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000156216.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156216.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008292.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008292.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008292.hg.1 |
ENST00000435367.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008292.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008292.hg.1 |
NR_120670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008292.hg.1 |
NR_120670.1 |
Refseq |