Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146278.hg.1 |
ENST00000424460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146278.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146278.hg.1 |
TC0700006688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146278.hg.1 |
NR_125740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146278.hg.1 |
ENST00000424460.1 |
Ensembl |