Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181951.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181951.hg.1 |
TC0500011889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181951.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181938.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181938.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181938.hg.1 |
TC0500011889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181938.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181938.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
TC0500011889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181945.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000519703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000520192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011889.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181939.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181939.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181939.hg.1 |
TC0500011889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181939.hg.1 |
ENST00000628324.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181939.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181949.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181949.hg.1 |
TC0500011889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181949.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181950.hg.1 |
NR_125774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181950.hg.1 |
TC0500011889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181950.hg.1 |
NR_125774.1 |
Refseq |