Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012920.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
TC0600012920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189248.hg.1 |
ENST00000452675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189257.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189257.hg.1 |
NR_125844.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189257.hg.1 |
TC0600012920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189257.hg.1 |
ENST00000427157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189257.hg.1 |
NR_125844.1 |
Refseq |