Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158839.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158839.hg.1 |
TC0600008466.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600158839.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158846.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158846.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158846.hg.1 |
TC0600008466.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600158846.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158846.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008466.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158837.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158837.hg.1 |
TC0600008466.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600158837.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158842.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158842.hg.1 |
NR_125857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158842.hg.1 |
TC0600008466.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600158842.hg.1 |
ENST00000418403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158842.hg.1 |
NR_125857.1 |
Refseq |