Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017787.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017787.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017787.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017787.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017787.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017787.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234139.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234139.hg.1 |
TC0100017787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100234139.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017787.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017787.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017787.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017787.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017787.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017787.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
TC0100017787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234138.hg.1 |
NR_125962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234140.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234140.hg.1 |
NR_125961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234140.hg.1 |
TC0100017787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100234140.hg.1 |
ENST00000451795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234140.hg.1 |
NR_125961.1 |
Refseq |