Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000625113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000625113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
ENST00000625113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213989.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213989.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213989.hg.1 |
TC0100015287.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213989.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213989.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
TC0100015287.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213990.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000625113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000625113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
ENST00000625113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015287.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
TC0100015287.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
ENST00000527660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213988.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213987.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213987.hg.1 |
NR_125963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213987.hg.1 |
TC0100015287.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213987.hg.1 |
ENST00000412270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213987.hg.1 |
NR_125963.1 |
Refseq |