Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006672.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006672.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006672.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006672.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006672.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145503.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145503.hg.1 |
TC1300006672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145503.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006672.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006672.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006672.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006672.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006672.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007396.hg.1 |
ENST00000550177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145505.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145505.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145505.hg.1 |
TC1300006672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145505.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145505.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145506.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145506.hg.1 |
NR_046547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145506.hg.1 |
TC1300006672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145506.hg.1 |
ENST00000440657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145506.hg.1 |
NR_046547.1 |
Refseq |