Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011088.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175135.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175135.hg.1 |
NR_102703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175135.hg.1 |
TC0X00011088.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175135.hg.1 |
ENST00000427671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175135.hg.1 |
NR_102703.1 |
Refseq |