Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANK1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102707.1 | 3 - 4 | 295 | 95 | ||
GenBank mRNA | AW080472.1 | - | 1 - 2 | 95 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fibronectin type III and ankyrin repeat domains 1 | FANK1 | HSD13 | Anti-sense | Chr. 10 - 125896539 - 126009599 on NCBI Build GRCh38 | |
Downstream | ADAM metallopeptidase domain 12 | ADAM12 |
ADAM12-OT1, CAR10, MCMP, MCMPMltna, MLTN, MLTNA |
39.5kb | Sense | Chr. 10 - 126012385 - 126388558 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |