Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009719.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009719.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009719.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009719.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166368.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166368.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166368.hg.1 |
TC1000009719.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000166368.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166368.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166370.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166370.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166370.hg.1 |
TC1000009719.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000166370.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166370.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009719.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009719.hg.1 |
ENST00000628989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009719.hg.1 |
NR_108074.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009719.hg.1 |
NR_108074.1 |
Refseq |