Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008333.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
TC0700008333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158157.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
TC0700008333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158151.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
TC0700008333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158152.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
TC0700008333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158158.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158156.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158156.hg.1 |
TC0700008333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158156.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000419668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000424523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000435695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
ENST00000452050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008333.hg.1 |
NR_110088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158155.hg.1 |
NR_110086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158155.hg.1 |
TC0700008333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158155.hg.1 |
NR_110086.1 |
Refseq |